Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: TNF655, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23472601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23472601

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Functional Assay
Format Purified
Cross Reactivity Human
Quantity 0.1mg
Gene ID (Entrez) 7124
Regulatory Status RUO
Isotype IgG1, κ
Primary or Secondary Primary
Antigen TNF-alpha
Host Species Mouse
Conjugate Unlabeled
Immunogen Recombinant human TNF-alpha
Gene Accession No. P01375
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone TNF655
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL