Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: SPM543, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437205

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Format Purified
Formulation PBS with 0.05% BSA.
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Zebrafish
Isotype IgG1, κ
Gene ID (Entrez) 7124
Reconstitution Protein A purified
Primary or Secondary Primary
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Clone SPM543
Purification Method Purified
Antigen TNF-alpha
Concentration 0.2mg/mL
Test Specificity TNF alpha exists as a multimer of two, three, or five non-covalently linked units, but shows a single 17kDa band following SDS PAGE under non-reducing conditions. TNF alpha is closely related to the 25kDa protein Tumor Necrosis Factor beta (lymphotoxin), sharing the same receptors and cellular actions.
Gene Accession No. P01375
Immunogen A hexadecapeptide corresponding to aa115-130 of human TNF-alpha
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Regulatory Status RUO
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.