Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: SPM543, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441902

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Conjugate Unlabeled
Quantity 0.2mg
Format Purified
Antigen TNF-alpha
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Zebrafish
Regulatory Status RUO
Host Species Mouse
Gene ID (Entrez) 7124
Concentration 1mg/mL
Gene Accession No. P01375
Primary or Secondary Primary
Purification Method Purified
Reconstitution Protein A purified
Immunogen A hexadecapeptide corresponding to aa115-130 of human TNF-alpha
Test Specificity TNF alpha exists as a multimer of two, three, or five non-covalently linked units, but shows a single 17kDa band following SDS PAGE under non-reducing conditions. TNF alpha is closely related to the 25kDa protein Tumor Necrosis Factor beta (lymphotoxin), sharing the same receptors and cellular actions.
Clone SPM543
Formulation 10mM PBS. Does not contain BSA. Azide Free.