Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: SPM543, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441901

  • / Each
 

Description & Specifications

Specifications

Immunogen A hexadecapeptide corresponding to aa115-130 of human TNF-alpha
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Zebrafish
Purification Method Purified
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Regulatory Status RUO
Antigen TNF-alpha
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01375
Host Species Mouse
Isotype IgG1, κ
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7124
Test Specificity TNF alpha exists as a multimer of two, three, or five non-covalently linked units, but shows a single 17kDa band following SDS PAGE under non-reducing conditions. TNF alpha is closely related to the 25kDa protein Tumor Necrosis Factor beta (lymphotoxin), sharing the same receptors and cellular actions.
Reconstitution Protein A purified
Clone SPM543
Concentration 1mg/mL
Conjugate Unlabeled