Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: P/T2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23430205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23430205

  • / Each
 

Description & Specifications

Specifications

Antigen TNF-alpha
Format Purified
Host Species Mouse
Immunogen A hexadecapeptide corresponding to aa115-130 of human TNF-alpha
Isotype IgG1, κ
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Zebrafish
Formulation PBS with 0.05% BSA.
Gene Accession No. P01375
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Purification Method Purified
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 7124
Reconstitution Protein A purified
Clone P/T2
Concentration 0.2mg/mL
Conjugate Unlabeled