Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: P/T2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23430201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23430201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Zebrafish
Antigen TNF-alpha
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01375
Immunogen A hexadecapeptide corresponding to aa115-130 of human TNF-alpha
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7124
Reconstitution Protein A purified
Clone P/T2
Concentration 0.2mg/mL
Conjugate Unlabeled
Host Species Mouse