Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: P/T2, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470301

  • / Each
 

Description & Specifications

Specifications

Antigen TNF-alpha
Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Zebrafish
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01375
Immunogen A hexadecapeptide corresponding to aa115-130 of human TNF-alpha
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7124
Reconstitution Protein A purified
Clone P/T2
Concentration 1mg/mL