Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: P/T2, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470302

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Primary or Secondary Primary
Regulatory Status RUO
Clone P/T2
Gene Accession No. P01375
Host Species Mouse
Concentration 1mg/mL
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Zebrafish
Quantity 0.2mg
Gene ID (Entrez) 7124
Immunogen A hexadecapeptide corresponding to aa115-130 of human TNF-alpha
Isotype IgG1, κ
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Antigen TNF-alpha
Purification Method Purified