Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: MAb1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23430501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23430501

  • / Each
 

Description & Specifications

Specifications

Antigen TNF-alpha
Gene Accession No. P01375
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human TNF-alpha
Isotype IgG1, κ
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7124
Clone MAb1
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Quantity 0.1mL
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Concentration 0.2mg/mL
Conjugate Unlabeled
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.