Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: MAb1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen TNF-alpha
Clone MAb1
Concentration 1mg/mL
Immunogen Recombinant human TNF-alpha
Gene Accession No. P01375
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23468201

 
novus biologicals canada
NBP23468201MG
0.1mg Each for $384.00

NBP23468202

 
novus biologicals canada
NBP23468202MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen TNF-alpha
Clone MAb1
Concentration 1mg/mL
Immunogen Recombinant human TNF-alpha
Gene Accession No. P01375
Gene ID (Entrez) 7124
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blotting