Please login to your online account to display your discounted pricing

Mouse anti-TNF-alpha, Clone: MAb1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23468201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23468201

  • / Each
 

Description & Specifications

Specifications

Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Neutralization
Applications Western Blotting
Cross Reactivity Human
Antigen TNF-alpha
Host Species Mouse
Immunogen Recombinant human TNF-alpha
Regulatory Status RUO
Gene ID (Entrez) 7124
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01375
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone MAb1
Concentration 1mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.