Please login to your online account to display your discounted pricing

Mouse anti-TLR2, Clone: mT2.7, Biotin, Novus Biologicals™

$131.00 - $362.00

Specifications

Antigen TLR2
Clone mT2.7
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was Tolllike receptor 2.
Gene ID (Entrez) 7097
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14348201

 
novus biologicals canada
NBP14348201MG
0.1mg Each for $362.00

NBP14348203

 
novus biologicals canada
NBP1434820025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen TLR2
Clone mT2.7
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was Tolllike receptor 2.
Gene ID (Entrez) 7097
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry