Please login to your online account to display your discounted pricing

Mouse anti-TLR2, Clone: 67N8F8, FITC, Novus Biologicals™

$128.00 - $406.00

Specifications

Antigen TLR2
Clone 67N8F8
Immunogen Within the range of amino acids 227-398 of human TLR2 were used as the immunogen for the antibody.
Gene Accession No. NP_003255.2
Gene ID (Entrez) 7097
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP231075

 
novus biologicals canada
NBP231075100TESTS
100 Tests Each for $406.00

NBP23107525

 
novus biologicals canada
NBP23107525TESTS
25 Tests Each for $128.00
Description & Specifications

Specifications

Antigen TLR2
Clone 67N8F8
Immunogen Within the range of amino acids 227-398 of human TLR2 were used as the immunogen for the antibody.
Gene Accession No. NP_003255.2
Gene ID (Entrez) 7097
Formulation TLR2 antibody at 5μL/test in 10mM Tris, 150mM NaCl. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate FITC
Format Purified
Purification Method Purified
Applications Flow Cytometry