Please login to your online account to display your discounted pricing

Mouse anti-TLR2, Clone: 67N8F8, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23107525TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23107525

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Quantity 25 Tests
Cross Reactivity Human
Format Purified
Gene Accession No. NP_003255.2
Host Species Mouse
Immunogen Within the range of amino acids 227-398 of human TLR2 were used as the immunogen for the antibody.
Isotype IgG1, κ
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7097
Clone 67N8F8
Conjugate FITC
Antigen TLR2
Formulation TLR2 antibody at 5μL/test in 10mM Tris, 150mM NaCl. Protein G purified.
Purification Method Purified
Storage Requirements Store at 4°C in the dark.
Monoclonal or Polyclonal Monoclonal