Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: TGB04+TGB05, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23429505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23429505

  • / Each
 

Description & Specifications

Specifications

Antigen Thyroglobulin
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone TGB04+TGB05
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01266
Host Species Mouse
Immunogen Human thyroid follicular cells
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7038
Reconstitution Protein A purified