Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: TGB04+TGB05, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Thyroglobulin
Clone TGB04+TGB05
Concentration 1mg/mL
Immunogen Human thyroid follicular cells
Gene Accession No. P01266
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23472401

 
novus biologicals canada
NBP23472401MG
0.1mg Each for $384.00

NBP23472402

 
novus biologicals canada
NBP23472402MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Thyroglobulin
Clone TGB04+TGB05
Concentration 1mg/mL
Immunogen Human thyroid follicular cells
Gene Accession No. P01266
Gene ID (Entrez) 7038
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot