Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: TGB04+TGB05, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23472401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23472401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Thyroglobulin
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01266
Host Species Mouse
Immunogen Human thyroid follicular cells
Isotype IgG1, κ
Quantity 0.1mg
Primary or Secondary Primary
Gene ID (Entrez) 7038
Reconstitution Protein A purified
Clone TGB04+TGB05
Concentration 1mg/mL
Conjugate Unlabeled
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.