Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: SPM517, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23299701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23299701

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mL
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01266
Immunogen Human thyroid follicular cells
Purification Method Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Thyroglobulin
Host Species Mouse
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 7038
Reconstitution Protein A purified
Clone SPM517
Concentration 0.2mg/mL