Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: SPM517, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Thyroglobulin
Clone SPM517
Concentration 1mg/mL
Immunogen Human thyroid follicular cells
Gene Accession No. P01266
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23477701

 
novus biologicals canada
NBP23477701MG
0.1mg Each for $384.00

NBP23477702

 
novus biologicals canada
NBP23477702MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Thyroglobulin
Clone SPM517
Concentration 1mg/mL
Immunogen Human thyroid follicular cells
Gene Accession No. P01266
Gene ID (Entrez) 7038
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot