Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: SPM517, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23477701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23477701

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Conjugate Unlabeled
Gene ID (Entrez) 7038
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Thyroglobulin
Concentration 1mg/mL
Host Species Mouse
Quantity 0.1mg
Clone SPM517
Purification Method Purified
Gene Accession No. P01266
Format Purified
Reconstitution Protein A purified
Immunogen Human thyroid follicular cells
Regulatory Status RUO
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.