Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: SPM517, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23477702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23477702

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Clone SPM517
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Conjugate Unlabeled
Quantity 0.2mg
Immunogen Human thyroid follicular cells
Gene ID (Entrez) 7038
Antigen Thyroglobulin
Reconstitution Protein A purified
Host Species Mouse
Purification Method Purified
Gene Accession No. P01266
Isotype IgG1, κ
Concentration 1mg/mL