Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: SPM221, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Thyroglobulin
Clone SPM221
Concentration 0.2mg/mL
Immunogen Human thyroid follicular cells
Gene Accession No. P01266
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23290410

 
novus biologicals canada
NBP23290410ML
1.0mL Each for $429.00

NBP23290401

 
novus biologicals canada
NBP23290401ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen Thyroglobulin
Clone SPM221
Concentration 0.2mg/mL
Immunogen Human thyroid follicular cells
Gene Accession No. P01266
Gene ID (Entrez) 7038
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting