Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: SPM221, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23290410ML

 View all options for this product


For Research Use Only

Catalog No. NBP23290410


Description & Specifications


Antigen Thyroglobulin
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone SPM221
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01266
Host Species Mouse
Immunogen Human thyroid follicular cells
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7038
Reconstitution Protein A purified