Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: SPM221, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23290401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23290401

  • / Each
 

Description & Specifications

Specifications

Antigen Thyroglobulin
Gene Accession No. P01266
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Purification Method Purified
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Human thyroid follicular cells
Isotype IgG1, κ
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7038
Reconstitution Protein A purified
Clone SPM221
Concentration 0.2mg/mL