Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: SPM221, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474801

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Purification Method Purified
Gene ID (Entrez) 7038
Quantity 0.1mg
Primary or Secondary Primary
Concentration 1mg/mL
Clone SPM221
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Antigen Thyroglobulin
Immunogen Human thyroid follicular cells
Reconstitution Protein A purified
Monoclonal or Polyclonal Monoclonal
Format Purified
Regulatory Status RUO
Gene Accession No. P01266
Host Species Mouse