Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: 2H11+6E1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23429401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23429401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Host Species Mouse
Antigen Thyroglobulin
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Clone 2H11+6E1
Gene ID (Entrez) 7038
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Gene Accession No. P01266
Concentration 0.2mg/mL
Purification Method Purified
Isotype IgG1, κ
Immunogen Human thyroid follicular cells