missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: 2H11+6E1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Thyroglobulin
Clone 2H11+6E1
Concentration 1mg/mL
Immunogen Human thyroid follicular cells
Gene Accession No. P01266
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23453001 Novus Biologicals Canada
NBP23453001MG
0.1mg Each for $384.00
NBP23453002 Novus Biologicals Canada
NBP23453002MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Thyroglobulin
1mg/mL
P01266
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
2H11+6E1
Human thyroid follicular cells
7038
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow Cytometry,Immunohistochemistry (Paraffin),Western Blot