Please login to your online account to display your discounted pricing

Mouse anti-Thyroglobulin, Clone: 2H11+6E1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23453002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23453002

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Thyroglobulin
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Immunogen Human thyroid follicular cells
Regulatory Status RUO
Gene Accession No. P01266
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7038
Quantity 0.2mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG1, κ
Concentration 1mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Purification Method Purified
Format Purified
Conjugate Unlabeled
Host Species Mouse
Clone 2H11+6E1