Please login to your online account to display your discounted pricing

Mouse anti-Thymidylate Synthase, Clone: TS106 + TMS715, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444705

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Antigen Thymidylate Synthase
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Recombinant human thymidylate synthase
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7298
Conjugate Unlabeled
Gene Accession No. P04818
Reconstitution Protein A purified
Clone TS106 + TMS715
Concentration 0.2mg/mL