missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Thymidylate Synthase, Clone: TS106 + TMS715, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Thymidylate Synthase
Clone TS106 + TMS715
Concentration 1mg/mL
Immunogen Recombinant human thymidylate synthase
Gene Accession No. P04818
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23450301 Novus Biologicals Canada
NBP23450301MG
0.1mg Each for $384.00
NBP23450302 Novus Biologicals Canada
NBP23450302MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Thymidylate Synthase
1mg/mL
P04818
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
TS106 + TMS715
Recombinant human thymidylate synthase
7298
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot