Please login to your online account to display your discounted pricing

Mouse anti-Thymidylate Synthase, Clone: TS106, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444505

  • / Each
 

Description & Specifications

Specifications

Format Purified
Cross Reactivity Human
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 7298
Reconstitution Protein A purified
Conjugate Unlabeled
Antigen Thymidylate Synthase
Formulation PBS with 0.05% BSA.
Gene Accession No. P04818
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human thymidylate synthase
Isotype IgG1, κ
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Clone TS106
Concentration 0.2mg/mL