Please login to your online account to display your discounted pricing

Mouse anti-Thymidylate Synthase, Clone: TS106, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Thymidylate Synthase
Clone TS106
Concentration 1mg/mL
Immunogen Recombinant human thymidylate synthase
Gene Accession No. P04818
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23450201

 
novus biologicals canada
NBP23450201MG
0.1mg Each for $384.00

NBP23450202

 
novus biologicals canada
NBP23450202MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Thymidylate Synthase
Clone TS106
Concentration 1mg/mL
Immunogen Recombinant human thymidylate synthase
Gene Accession No. P04818
Gene ID (Entrez) 7298
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot