Please login to your online account to display your discounted pricing

Mouse anti-Thymidylate Synthase, Clone: TS106, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23450201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23450201

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Thymidylate Synthase
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Gene Accession No. P04818
Host Species Mouse
Immunogen Recombinant human thymidylate synthase
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Primary or Secondary Primary
Gene ID (Entrez) 7298
Reconstitution Protein A purified
Clone TS106
Cross Reactivity Human