Please login to your online account to display your discounted pricing

Mouse anti-Thymidylate Synthase, Clone: TMS715, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23472501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23472501

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Thymidylate Synthase
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Recombinant human thymidylate synthase
Isotype IgG1, κ
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7298
Reconstitution Protein A purified
Conjugate Unlabeled
Gene Accession No. P04818
Host Species Mouse
Purification Method Purified
Primary or Secondary Primary
Clone TMS715
Concentration 1mg/mL
Cross Reactivity Human