Please login to your online account to display your discounted pricing

Mouse anti-Thymidylate Synthase, Clone: SPM453, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Thymidylate Synthase
Clone SPM453
Concentration 0.2mg/mL
Immunogen Recombinant human thymidylate synthase
Gene Accession No. P04818
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23439001

 
novus biologicals canada
NBP23439001ML
0.1mL Each for $139.00

NBP23439005

 
novus biologicals canada
NBP23439005ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Thymidylate Synthase
Clone SPM453
Concentration 0.2mg/mL
Immunogen Recombinant human thymidylate synthase
Gene Accession No. P04818
Gene ID (Entrez) 7298
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting