Please login to your online account to display your discounted pricing

Mouse anti-Thymidylate Synthase, Clone: SPM453, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439001

  • / Each
 

Description & Specifications

Specifications

Antigen Thymidylate Synthase
Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human thymidylate synthase
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity It recognizes a protein of 36kDa, identified as Thymidylate Synthase (TS) (EC 2.1.1.45).
Reconstitution Protein A purified
Conjugate Unlabeled
Gene Accession No. P04818
Isotype IgG1, κ
Quantity 0.1mL
Gene ID (Entrez) 7298
Clone SPM453
Cross Reactivity Human
Format Purified
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 0.2mg/mL