Please login to your online account to display your discounted pricing

Mouse anti-Thymidylate Synthase, Clone: SPM453, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444101

  • / Each
 

Description & Specifications

Specifications

Antigen Thymidylate Synthase
Immunogen Recombinant human thymidylate synthase
Isotype IgG1, κ
Conjugate Unlabeled
Gene Accession No. P04818
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Clone SPM453
Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7298
Purification Method Purified
Test Specificity It recognizes a protein of 36kDa, identified as Thymidylate Synthase (TS) (EC 2.1.1.45).
Reconstitution Protein A purified
Concentration 1mg/mL