Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: TG86+P/T1, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen TGF-alpha
Clone TG86+P/T1
Concentration 0.2mg/mL
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGFalpha (TG86 and P/T1)
Gene Accession No. P01135
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23430001

 
novus biologicals canada
NBP23430001ML
0.1mL Each for $139.00

NBP23430005

 
novus biologicals canada
NBP23430005ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen TGF-alpha
Clone TG86+P/T1
Concentration 0.2mg/mL
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGFalpha (TG86 and P/T1)
Gene Accession No. P01135
Gene ID (Entrez) 7039
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Zebrafish
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation