Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: TG86+P/T1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23472301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23472301

  • / Each
 

Description & Specifications

Specifications

Antigen TGF-alpha
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Clone TG86+P/T1
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01135
Host Species Murine
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGFalpha (TG86 and P/T1)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7039
Test Specificity This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody.
Reconstitution Protein A purified
Target Species Human
Target Species Rabbit
Target Species Zebrafish