Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: TG86, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23472201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23472201

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01135
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGFalpha.
Isotype IgG1, κ
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody.
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Zebrafish
Antigen TGF-alpha
Format Purified
Host Species Mouse
Purification Method Purified
Regulatory Status RUO
Gene ID (Entrez) 7039
Reconstitution Protein A purified
Clone TG86