Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: SPM542, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437105

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 7039
Test Specificity This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin.
Reconstitution Protein A purified
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Zebrafish
Antigen TGF-alpha
Gene Accession No. P01135
Host Species Mouse
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGF alpha
Monoclonal or Polyclonal Monoclonal
Clone SPM542
Concentration 0.2mg/mL
Conjugate Unlabeled