Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: SPM542, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen TGF-alpha
Clone SPM542
Concentration 1mg/mL
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGF alpha
Gene Accession No. P01135
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23441801

 
novus biologicals canada
NBP23441801MG
0.1mg Each for $384.00

NBP23441802

 
novus biologicals canada
NBP23441802MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen TGF-alpha
Clone SPM542
Concentration 1mg/mL
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGF alpha
Gene Accession No. P01135
Gene ID (Entrez) 7039
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation