Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: SPM542, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441802

  • / Each
 

Description & Specifications

Specifications

Antigen TGF-alpha
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGF alpha
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7039
Test Specificity This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin.
Clone SPM542
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Zebrafish
Gene Accession No. P01135
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Reconstitution Protein A purified
Concentration 1mg/mL