Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: SPM542, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441801

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Quantity 0.1mg
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGF alpha
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Primary or Secondary Primary
Test Specificity This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Concentration 1mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen TGF-alpha
Clone SPM542
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Zebrafish
Regulatory Status RUO
Gene ID (Entrez) 7039
Reconstitution Protein A purified
Host Species Mouse
Purification Method Purified
Gene Accession No. P01135
Isotype IgG1, κ