Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: P/T1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470202

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGFalpha.
Monoclonal or Polyclonal Monoclonal
Quantity 0.2mg
Gene Accession No. P01135
Conjugate Unlabeled
Purification Method Purified
Gene ID (Entrez) 7039
Regulatory Status RUO
Test Specificity This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Clone P/T1
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Zebrafish
Isotype IgG1, κ
Concentration 1mg/mL
Antigen TGF-alpha
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Format Purified
Reconstitution Protein A purified
Primary or Secondary Primary