Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: MF9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23429605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23429605

  • / Each
 

Description & Specifications

Specifications

Immunogen A 17-amino acid synthetic peptide from the carboxyl-terminus of rat TGFalpha.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Conjugate Unlabeled
Format Purified
Formulation PBS with 0.05% BSA.
Clone MF9
Gene ID (Entrez) 7039
Quantity 0.5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Isotype IgG1, κ
Reconstitution Protein A purified
Gene Accession No. P01135
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 0.2mg/mL
Regulatory Status RUO
Purification Method Purified
Antigen TGF-alpha