Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: MF9, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23468302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23468302

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P01135
Concentration 1mg/mL
Purification Method Purified
Immunogen A 17-amino acid synthetic peptide from the carboxyl-terminus of rat TGFalpha.
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Clone MF9
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Format Purified
Gene ID (Entrez) 7039
Isotype IgG1, κ
Host Species Mouse
Reconstitution Protein A purified
Antigen TGF-alpha
Quantity 0.2mg