Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: 1E8-G6, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen TGF-alpha
Clone 1E8-G6
Concentration 0.2mg/mL
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGFalpha.
Gene Accession No. P01135
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23429701

 
novus biologicals canada
NBP23429701ML
0.1mL Each for $139.00

NBP23429705

 
novus biologicals canada
NBP23429705ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen TGF-alpha
Clone 1E8-G6
Concentration 0.2mg/mL
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGFalpha.
Gene Accession No. P01135
Gene ID (Entrez) 7039
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Zebrafish
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation