Please login to your online account to display your discounted pricing

Mouse anti-TGF-alpha, Clone: 1E8-G6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23429705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23429705

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Rabbit
Cross Reactivity Zebrafish
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 7039
Antigen TGF-alpha
Gene Accession No. P01135
Quantity 0.5mL
Format Purified
Immunogen A 10-amino acid synthetic peptide (aa34-43) from human TGFalpha.
Monoclonal or Polyclonal Monoclonal
Clone 1E8-G6
Reconstitution Protein A purified
Concentration 0.2mg/mL
Regulatory Status RUO
Host Species Mouse
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody.
Isotype IgG1, κ
Primary or Secondary Primary
Purification Method Purified