Please login to your online account to display your discounted pricing

Mouse anti-TfR (Transferrin R), Clone: DF1513, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23460202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23460202

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Host Species Mouse
Quantity 0.2mg
Regulatory Status RUO
Cross Reactivity Human
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone DF1513
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Primary or Secondary Primary
Antigen TfR (Transferrin R)
Conjugate Unlabeled
Immunogen KG1 acute myeloid leukemia cell line
Gene Accession No. P02786
Gene ID (Entrez) 7037
Reconstitution Protein A purified
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal
Purification Method Purified